storage temp.
−20°C
SMILES string
[H][C@@](CC(C)C)(NC(C)=O)C(O)=O
InChI
1S/C8H15NO3/c1-5(2)4-7(8(11)12)9-6(3)10/h5,7H,4H2,1-3H3,(H,9,10)(H,11,12)
InChI key
WXNXCEHXYPACJF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
BMC neurology, 17(1), 7-7 (2017-01-11)
Cerebellar ataxia (CA) is a frequent and often disabling condition that impairs motor functioning and impacts on quality of life (QoL). No medication has yet been proven effective for the symptomatic or even causative treatment of hereditary or non-hereditary, non-acquired
PloS one, 15(2), e0229585-e0229585 (2020-02-29)
The enantiomers of many chiral drugs not only exhibit different pharmacological effects in regard to targets that dictate therapeutic and toxic effects, but are also handled differently in the body due to pharmacokinetic effects. We investigated the pharmacokinetics of the
Journal of chromatography. A, 1583, 80-87 (2018-11-26)
Modified metabolites play important roles in diagnostic monitoring, oxidative response and physiological regulation. Comprehensive analytical methods are greatly needed for improving the coverage of modified metabolites and studying their physiological function. Here, a novel nontargeted profiling method for mapping modified
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service