Skip to Content
Merck
All Photos(1)

Key Documents

97596

Sigma-Aldrich

1,2,3-Octanetriol Isomer T (74888) 1 M solution

Synonym(s):

1,2,3-Octanetriol solution, Additive Screening Solution 11/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

CCCCCC(O)C(O)CO

InChI

1S/C8H18O3/c1-2-3-4-5-7(10)8(11)6-9/h7-11H,2-6H2,1H3

InChI key

QSQGNBGEHSFMAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service