Skip to Content
Merck
All Photos(1)

Key Documents

89945

Sigma-Aldrich

1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine, 7-nitrobenzofurazan-labeled

BioReagent, suitable for fluorescence, ammonium salt, ≥98.0% (TLC)

Synonym(s):

L-β,γ-Dimyristoyl-α-cephaline, 7-nitrobenzofurazan-labeled, N-(7-Nitro-benzofurazan-4-yl)-1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine, Phosphatidylethanolamine 14:0, NBD-labeled

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H67N4O11P
CAS Number:
Molecular Weight:
798.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

Assay

≥98.0% (TLC)

fluorescence

λex 459 nm; λem 533 nm

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OCCNc1ccc([N+]([O-])=O)c2nonc12)OC(=O)CCCCCCCCCCCCC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service