80074
Ammonium sulfate 0.17M – Sodium cacodylate pH 6.5; 0.085M – Polyethylene glycol 8000 25.5% (w/v) – Glycerol 15% (v/v) solution
Synonym(s):
Crystallization solution 15/Kit No 75403
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
form
liquid
pH
6.5±0.2 ( in neat)
density
1.10 g/mL at 20 °C
storage temp.
2-8°C
Signal Word
Danger
Hazard Statements
Precautionary Statements
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service