Skip to Content
Merck
All Photos(1)

Key Documents

76724

Sigma-Aldrich

meso-Tetraphenyl-tetrabenzoporphine Palladium Complex

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H36N4Pd
CAS Number:
Molecular Weight:
919.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0%

storage temp.

2-8°C

SMILES string

[Pd]1n2c3c(-c4ccccc4)c5nc(c(-c6ccccc6)c7n1c(c(-c8ccccc8)c9nc(c(-c%10ccccc%10)c2c%11ccccc3%11)c%12ccccc9%12)c%13ccccc7%13)c%14ccccc5%14

Application

Phosphorescent probe for oxygen measurements in very low concentration

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.A. Vinogradov et al.
J. Chem. Soc. Perkin Trans. II, 103-103 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service