Skip to Content
Merck
All Photos(1)

Key Documents

62350

Sigma-Aldrich

Lipozyme®, immobilized from Mucor miehei

granular, brown, >30 U/g

Synonym(s):

Lipase, immobilized on a macroporous ion-exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

granular

specific activity

>30 U/g

color

brown

storage temp.

2-8°C

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Unit Definition

1 U corresponds to the amount of enzyme which sets free 1 μmol stearic acid per minute at pH 8.0 and 70 °C (tristearin, Cat. No. 69498 as substrate)

Other Notes

A 1,3-specific lipase for interesterification and ester synthesis

Legal Information

Lipozyme is a registered trademark of Novo Nordisk A/S Corp.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Nicolosi et al.
Tetrahedron Letters, 36, 6545-6545 (1995)
T.T. Hansen et al.
Proc.- World Conf. Emerging Technol. Fats Oils Ind. Ed. B.A. Richard, 365-365 (1985)
Guan-Chun Chen et al.
Biotechnology letters, 33(3), 525-529 (2010-11-04)
Triacylglycerol lipase from Pseudomonas cepacia and Fe(3)O(4) magnetic nanoparticles were encapsulated simultaneously within biomimetic silica through the catalysis of polyallylamine. The encapsulation efficiency reached 96% with an activity recovery of 51%. After 5 h at 37°C, the activities of the
Industrial applications of microbial lipases
F. Hasan et al.
Enzyme and Microbial Technology, 39, 235-251 (2006)
B Sànchez-Nogué et al.
Environmental science and pollution research international, 20(5), 3480-3488 (2012-12-06)
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service