Skip to Content
Merck
All Photos(1)

Key Documents

56902C

SAFC

Grace′s Insect Medium Original Formula

With L-glutamine, dry powder

Synonym(s):

Grace’s Insect Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

description

for research or for further manufacturing use
with 4.1 mM L-glutamine, without calcium chloride, without sodium bicarbonate

form

dry powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Grace′s medium was originally formulated to support the growth of cells derived from the Australian emperor gum moth, Antherea eucalypti. The medium is a modification of Wyatt′s medium to more closely resemble Antherea hemolymph. The cell lines established by Grace using this medium were the first continuous lines developed. The basal medium, when properly supplemented, has been used to culture cells derived from a variety of insects including several species of lepidopterans as well as some dipterans. The medium is primarily used as a basal medium for the growth and maintenance of cell lines derived from lepidopterans.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service