Skip to Content
Merck
All Photos(1)

Documents

54375

Supelco

Micro particles based on silicon dioxide

size: 4 μm

Synonym(s):

Silica dioxide microsphere (4 μm), Micro silicon (4 μm), Beads based on silicon dioxide, microsize, Silicon dioxide beads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

grade

analytical standard

form

aqueous suspension

concentration

5% (solids)

particle size

4 μm std dev ≤0.3 μm

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silicon dioxide based microparticles are ideal to obtain particle size distribution (PSD) profile of test samples.
The particle diameter of 4 μm is determined using Coulter Multisizer.

Application

Used to calibrate, qualify and check the performance of particle size analyzers, surface scanning instruments and particle counters.
Also used to prepare working fluid standard for use with a microfluidic particle concentrator.

Features and Benefits

  • suitable for routine instrument calibration monitoring, testing and corrections
  • particle size traceable to Community Bureau of Reference (BCR) standards
  • available in 5 mL pack size as a neat sample

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scaled particle focusing in a microfluidic device with asymmetric electrodes utilizing induced-charge electroosmosis
Ren Y, et al.
Lab on a chip, 16(15), 2803-2812 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service