Skip to Content
Merck
All Photos(1)

Key Documents

47753

Sigma-Aldrich

Formate Dehydrogenase from Candida boidinii

liquid (clear), clear brown, 40.0-60.0 U/mL

Synonym(s):

FDH, Formate:NAD+ oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

biological source

fungus (Candida boidinii)

form

liquid (clear)

mol wt

Mr ~76000

concentration

40.0-60.0 U/mL

color

clear brown

density

1.1 g/mL at 20 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Formate dehydrogenase is involved in the stress response of plants and catalyzes the reduction of NAD+ to NADH.

Unit Definition

1 U corresponds to the amount of enzyme which oxidizes 1 μmol sodium formate (Cat. No. 71539) per minute at pH 7.6 and 25 °C

Other Notes

Preferred enzyme for regenerating NADH from NAD

comparable product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Drauz et al.
Enzyme Catalysis in Organic Synthesis, 597-597 (1995)
Frances L Shaw et al.
Journal of bacteriology, 194(15), 3814-3823 (2012-05-23)
The food-borne bacterial pathogen Campylobacter jejuni efficiently utilizes organic acids such as lactate and formate for energy production. Formate is rapidly metabolized via the activity of the multisubunit formate dehydrogenase (FDH) enzyme, of which the FdhA subunit is predicted to
Yutaka Amao et al.
Faraday discussions, 155, 289-296 (2012-04-05)
Solar fuels, such as hydrogen gas produced from water and methanol produced from carbon dioxide reduction by artificial photosynthesis, have received considerable attention. In natural leaves the photosynthetic proteins are well-organized in the thylakoid membrane. To develop an artificial leaf
V I Tishkov et al.
Biochemistry. Biokhimiia, 69(11), 1252-1267 (2005-01-04)
NAD+-dependent formate dehydrogenase (FDH) is an abundant enzyme that plays an important role in energy supply of methylotrophic microorganisms and in response to stress in plants. FDH belongs to the superfamily of D-specific 2-hydroxy acid dehydrogenases. FDH is widely accepted
Yuan Lu et al.
Applied biochemistry and biotechnology, 167(4), 732-742 (2012-05-18)
To improve the hydrogen productivity and examine the hydrogen evolution mechanism of Clostridium paraputrificum, roles of formate in hydrogen evolution and effects of introducing formate-originated NADH regeneration were explored. The formate-decomposing pathway for hydrogen production was verified to exist in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service