Skip to Content
Merck
All Photos(1)

Key Documents

4370280

Sigma-Aldrich

iTRAQ® Reagent Application Kit - Plasma

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CHF 1’680.00

CHF 1’680.00


Check Cart for Availability


Select a Size

Change View
1 KIT
CHF 1’680.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.24

CHF 1’680.00


Check Cart for Availability

shipped in

dry ice

storage temp.

−20°C

General description

Contains sufficient reaction solutions and iTRAQ® Reagents 114, 115, 116 and 117 for 8 duplex or four 4-plex experiments based on 50 μg of protein per sample.

Application

This iTRAQ application kit for plasma provides iTRAQ reagents and protocol targeted specifically for plasma workflows. Cysteine blocking reagent is not included. Trypsin is sold separately.

Analysis Note

To view the Protocol for the iTRAQ Reagents Application Kit - Plasma, please visit this Protocol link.

Legal Information

iTraq is a registered trademark of AB Sciex Pte. Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service