Skip to Content
Merck
All Photos(1)

Key Documents

4339035

Sigma-Aldrich

Cleavable iCAT® Reagent - Methods Developments Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

General description

Cleavable ICAT reagents Methods Development Kit contains 3 units of Heavy reagent and 6 units of Light reagent. Each unit of reagent can label up to ~100 μg of protein.

Analysis Note

To view the Protocol for the Cleavable ICAT Reagent Kit for Protein Labeling (monoplex version), please visit this Protocol link

To view the Protocol for the 1-D PAGE Cleavable ICAT Reagent Applications Development Kit for Targeted Protein ID and Quantitation (monoplex version), please visit this Protocol link.

To view the Quick Reference Procedure for the 1-D PAGE Cleavable ICAT Reagent Applications Development Kit for Targeted Protein ID and Quantitation (monoplex version), please visit this Quick Reference Procedure link.

To view the Quick Reference Card for the Cleavable ICAT Reagent Kit for Protein Labeling, please visit this Quick Reference Card link.

Legal Information

iCAT is a registered trademark of University of Washington

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trairak Pisitkun et al.
Physiological genomics, 25(2), 263-276 (2006-02-02)
The inner medullary collecting duct (IMCD) is an important site of vasopressin-regulated water and urea transport. Here we have used protein mass spectrometry to investigate the proteome of the IMCD cell and how it is altered in response to long-term

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service