Skip to Content
Merck
All Photos(1)

Documents

40357

Sigma-Aldrich

Dextran sulfate sodium salt from Leuconostoc spp.

low sulfate content

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201

form

powder

quality

low sulfate content

mol wt

Mw ~20,000

composition

sulfur content, 8-13%

solubility

water: soluble

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran.

Application

Dextran sulfate (Sigma) can be used to precipitate the lipoprotein HDL from human plasma samples.
Dextran sulfate is routinely used for the selective precipitation of lipoproteins, probe hybridization membrane-immobilized DNA, the release of DNA from DNA-histone complexes, and the inhibition of RNA binding to ribosomes.

Preparation Note

Sigma dextrans are derived from Leuconostoc mesenteroides.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Navab et al.
Journal of lipid research, 42(8), 1308-1317 (2001-08-03)
We have developed a novel and rapid cell-free assay of the ability of HDL to prevent the formation of or inactivate oxidized phospholipids. HDL was tested for its ability to inhibit the oxidation of LDL, or inhibit the oxidation of
Aaron H Rose et al.
Inflammatory bowel diseases, 21(9), 2005-2015 (2015-06-16)
An important role has emerged for calpain enzymes in regulating inflammation with one isoform, calpain-2, particularly important for macrophage activation. The goal of this study was to determine the therapeutic potential of a synthetic calpain-2 inhibitor, zLLY-CH2F, for colitis and

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service