Skip to Content
Merck
All Photos(1)

Documents

39890

Sigma-Aldrich

N,N-Dimethylcasein

BioReagent, suitable as Substrate for Proteolytic Enzymes

Synonym(s):

N,N-Dimethylated casein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.28

product name

N,N-Dimethylcasein, BioReagent, for spectrophotometric det. of enzyme activity

product line

BioReagent

quality

for spectrophotometric det. of enzyme activity

solubility

water: 1.0 mg/mL, clear, colorless

λ

0.1 % in H2O (TBS test)

UV absorption

λ: 340 nm Amax: ≤0.08

storage temp.

2-8°C

Application

Substrate for the assay of proteolytic enzymes. Much more sensitive than the caseinolytic method of Kunitz

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kunitz
The Journal of general physiology, 30(4), 291-310 (1947-03-20)
A study has been made of the general properties of crystalline soybean trypsin inhibitor. The soy inhibitor is a stable protein of the globulin type of a molecular weight of about 24,000. Its isoelectric point is at pH 4.5. It
The action of proteolytic enzymes on N,N-dimethyl proteins. Basis for a microassay for proteolytic enzymes.
Y Lin et al.
The Journal of biological chemistry, 244(3), 789-793 (1969-02-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service