Skip to Content
Merck
All Photos(1)

Documents

24530C

SAFC

EX-CELL® EBx GRO-I Serum-Free Medium for Embryonic Stem Cells

without L-glutamine, without phenol red, without sodium bicarbonate

Pharma Manufacturing

Synonym(s):

EX-CELL® EBx Serum-Free Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

description

Animal-component free
Animal-component-free, low protein
for research or for further manufacturing use

Quality Level

form

dry powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C
2-8°C

Looking for similar products? Visit Product Comparison Guide

Quantity

Formulated to contain 19.1 grams of powder per liter of medium.

Legal Information

EBx is a trademark of Vivalis SA
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service