Skip to Content
Merck
All Photos(1)

Key Documents

20780

Sigma-Aldrich

Butyrylcholine iodide

≥99.0% (AT)

Synonym(s):

(2-Hydroxyethyl)trimethylammonium iodide butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOCH2CH2N(I)(CH3)3
CAS Number:
Molecular Weight:
301.17
Beilstein:
3917649
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Quality Level

Assay

≥99.0% (AT)

form

powder

mp

85-89 °C

solubility

methanol: 1 g/10 mL, clear, colorless

storage temp.

2-8°C

SMILES string

[I-].CCCC(=O)OCC[N+](C)(C)C

InChI

1S/C9H20NO2.HI/c1-5-6-9(11)12-8-7-10(2,3)4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

GALNBQVDMJRFGJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for cholinesterase (EC 3.1.1.8); and acetylcholinesterase (EC 3.1.1.7)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R D O'Brien et al.
Biochimica et biophysica acta, 526(1), 129-134 (1978-09-11)
Comparisons were made of purified acetylcholinesterase from the heads of wild type house flies with a mutant form (which bound organophosphates and carbamates less tightly). Using 12 substrates and 6 quaternary inhibitors, the only substantial difference was that the Km
Nasiara Karim et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 96, 700-709 (2017-10-19)
Rosmarinus officinalis has long been known as the herb of remembrance. The present study was undertaken to investigate the anti-amnesic effects of nepitrin isolated from Rosmarinus officinalis using in-vivo models of Y-maze and novel object recognition test (NORT) along with
Monitoring of air and water for enzyme inhibitors.
L H Goodson et al.
Methods in enzymology, 44, 647-658 (1976-01-01)
Vibhor Gupta et al.
PloS one, 14(11), e0225188-e0225188 (2019-11-26)
Rare diseases defined by genetic mutations are classic targets for gene therapy. More recently, researchers expanded the use of gene therapy in non-clinical studies to infectious diseases through the delivery of vectorized antibodies to well-defined antigens. Here, we further extend
Cüneyt Çağlayan et al.
Journal of biochemical and molecular toxicology, 33(10), e22381-e22381 (2019-08-28)
In this study, it was demonstrated the ameliorative effect of zingerone (ZO) (25 and 50 mg/kg body weight) against vancomycin (VCM) (200 mg/kg body weight) administered to rats on some metabolic enzymes' activities in the lung, liver, kidney, and testis tissues of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service