Skip to Content
Merck
All Photos(1)

Key Documents

14533C

SAFC

EX-CELL® EBx PRO-II Serum-Free Media

Animal-component free, without L-glutamine, without phenol red, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Synonym(s):

EX-CELL® EBx Serum-Free Media

Sign Into View Organizational & Contract Pricing

Select a Size

1000 ML
CHF 120.00

CHF 120.00


Estimated to ship on09 April 2025



Select a Size

Change View
1000 ML
CHF 120.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

CHF 120.00


Estimated to ship on09 April 2025


Quality Level

description

Animal-component-free, low protein
for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

availability

not available in Japan

technique(s)

cell culture | mammalian: suitable

components

L-glutamine: no
phenol red: no

shipped in

wet ice

storage temp.

2-8°C
2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

EBx is a trademark of Vivalis SA
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service