Skip to Content
Merck
All Photos(1)

Key Documents

12266

Sigma-Aldrich

N-[4-(2-Benzimidazolyl)phenyl]maleimide

BioReagent, suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

BIPM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11N3O2
CAS Number:
Molecular Weight:
289.29
Beilstein:
755407
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

Assay

≥98.0% (HPLC)

form

solid

solubility

H2O: soluble
acetone: soluble

fluorescence

λex 315 nm; λem 365 nm (after derivatization with 2-mercaptoethanol)

suitability

suitable for fluorescence

SMILES string

O=C1C=CC(=O)N1c2ccc(cc2)-c3nc4ccccc4[nH]3

InChI

1S/C17H11N3O2/c21-15-9-10-16(22)20(15)12-7-5-11(6-8-12)17-18-13-3-1-2-4-14(13)19-17/h1-10H,(H,18,19)

InChI key

NZDOXVCRXDAVII-UHFFFAOYSA-N

Other Notes

Fluorescent probe in proteins, used for microfluorometry of thiol compounds (detection limit: 10-6 m thiol conc.); Review

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Taniguchi et al.
The Journal of biological chemistry, 258(11), 6927-6931 (1983-06-10)
The addition of ATP to (Na+,K+)-ATPase (EC 3.6.1.3) of pig kidney modified with N-[p-(2-benzimidazolyl)phenyl]maleimide induced three different modes of fluorescence change: positive, biphasic, and negative, which were observed, respectively, in the presence of Mg2+ with 16 or 160, 320 or
Robert J W Brewin et al.
Sensors (Basel, Switzerland), 19(4) (2019-03-01)
Two expanding areas of science and technology are citizen science and three-dimensional (3D) printing. Citizen science has a proven capability to generate reliable data and contribute to unexpected scientific discovery. It can put science into the hands of the citizens
K Taniguchi et al.
Journal of biochemistry, 109(2), 299-306 (1991-02-01)
Oligomycin reduced the fluorescence intensity of an N-(p-(2-benzimidazoly)phenyl) maleimide (BIPM) probe at Cys-964 of the alpha-chain of pig kidney Na+,K(+)-ATPase with increase in the concentration of Na+ with a Hill coefficient of nh = 0.77 with Kh = 231 mM.
S Inoue et al.
Japanese journal of pharmacology, 43(1), 107-111 (1987-01-01)
Trypsin treatment of N-[p-(2-benzimidazolyl)phenyl]maleimide modified enzyme caused a marked reduction in Na+,K+-ATPase activity and in the amount of the alpha-chain, which contains the phosphorylation and ouabain binding sites. However, these preparations retained nearly 90% of the ouabain binding capacity and
Fluorescent thiol reagents. V. Microfluorometry of thiol compounds with a fluorescent-labeled maleimide.
T Sekine et al.
Analytical biochemistry, 48(2), 557-568 (1972-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service