Skip to Content
Merck
All Photos(1)

Key Documents

0710000

Sigma-Aldrich

4-tert-Butylphenoxyacetic acid

≥98.0% (HPLC)

Synonym(s):

TAC acid, [4-(1,1-Dimethylethyl)phenoxy]-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4OCH2CO2H
CAS Number:
Molecular Weight:
208.25
Beilstein:
2368208
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (HPLC)

mp

96 °C (lit.)

SMILES string

CC(C)(C)c1ccc(OCC(O)=O)cc1

InChI

1S/C12H16O3/c1-12(2,3)9-4-6-10(7-5-9)15-8-11(13)14/h4-7H,8H2,1-3H3,(H,13,14)

InChI key

FBIGAJNVRFKBJL-UHFFFAOYSA-N

Application

Reagent for the TAC protective group in nucleosides[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N D Sinha et al.
Biochimie, 75(1-2), 13-23 (1993-01-01)
The use of tert-butylphenoxyacetyl for N-protection of nucleoside bases during oligonucleotide synthesis facilitates its removal under mild conditions. This protecting group reduces depurination of deoxyadenosine residues, and minimizes premature desilylation resulting in chain degradation during post-synthesis workup of synthetic RNA.
Katarzyna Przejczowska-Pomierny et al.
Chirality, 29(9), 500-511 (2017-07-06)
A direct fluorometric high-performance liquid chromatography (HPLC) method was developed and validated for the analysis of ibuprofen enantiomers in mouse plasma (100 μl) and tissues (brain, liver, kidneys) using liquid-liquid extraction and 4-tertbutylphenoxyacetic acid as an internal standard. Separation of enantiomers
K E Cole et al.
The Journal of experimental medicine, 187(12), 2009-2021 (1998-06-24)
Chemokines are essential mediators of normal leukocyte trafficking as well as of leukocyte recruitment during inflammation. We describe here a novel non-ELR CXC chemokine identified through sequence analysis of cDNAs derived from cytokine-activated primary human astrocytes. This novel chemokine, referred

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service