Skip to Content
Merck
All Photos(1)

Key Documents

71283

Sigma-Aldrich

Ammonium sodium phosphate dibasic tetrahydrate

puriss. p.a., ≥99.0% (NT)

Synonym(s):

sec-Sodium ammonium phosphate, Ammonium sodium hydrogen phosphate, Sodium ammonium hydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNH4HPO4 · 4H2O
CAS Number:
Molecular Weight:
209.07
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

puriss. p.a.

Assay

≥99.0% (NT)

form

crystals

solubility

water: soluble(lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤10 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Tl: ≤10 mg/kg
Zn: ≤5 mg/kg

SMILES string

[NH4+].[Na+].OP([O-])([O-])=O

InChI

1S/H3N.Na.H3O4P.4H2O/c;;1-5(2,3)4;;;;/h1H3;;(H3,1,2,3,4);4*1H2/q;+1;;;;;/p-1

InChI key

IQTQISLCLWJRPM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium sodium phosphate dibasic tetrahydrate (Ammonium sodium hydrogen phosphate) is a colorless ammonium salt. On heating, it undergoes decomposition to afford sodium polyphosphate.[1]

Application

Ammonium sodium phosphate dibasic tetrahydrate (Sodium ammonium phosphate tetrahydrate) may be used in the following studies:
  • To compose the growth medium during the biocatalytic synthesis of 4-fluorocatechol.[2]
  • Preparation of microcosmic salt beads, which are used in qualitative analysis.[1]

Other Notes

Reagent for the decomposition of barium sulfate in phosphate salt beads[3][4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
12.7 The Biocatalytic Synthesis of 4-Fluorocatechol from Fluorobenzene.
Whittall J and Sutton P.
Practical Methods for Biocatalysis and Biotransformations, 379-379 (2009)
Eagleson M.
Concise Encyclopedia Chemistry, 67-67 (1994)
Caryn Wadler et al.
Analytical biochemistry, 368(1), 17-23 (2007-07-03)
A new approach to determine in vivo pools of coenzyme A (CoA) and short chain acyl-CoA thioesters is reported. The metabolites released by extraction with trichloroacetic acid are recovered and quantitatively dephosphorylated by treatment with shrimp alkaline phosphatase. Following phosphatase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service