45760
Ethyl acetate
puriss. p.a., ACS reagent, ≥99.5% (GC)
Synonym(s):
EtOAc
About This Item
grade
ACS reagent
puriss. p.a.
vapor density
3 (20 °C, vs air)
vapor pressure
73 mmHg ( 20 °C)
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
801 °F
expl. lim.
2.2-11.5 %, 38 °F
technique(s)
GC/GC: suitable
impurities
substances darkened by H2SO4, in accordance
≤0.005% free acid (as CH3COOH)
≤0.05% water
≤0.1% 2-methyl-1-propanol (GC)
≤0.1% ethanol (GC)
≤0.1% methanol (GC)
≤0.1% methyl acetate (GC)
evapn. residue
≤0.001%
refractive index
n20/D 1.3720 (lit.)
n20/D 1.372
bp
76.5-77.5 °C (lit.)
mp
−84 °C (lit.)
density
0.902 g/mL at 25 °C (lit.)
cation traces
Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg
SMILES string
CCOC(C)=O
format
neat
InChI
1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3
InChI key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- 3-(pyridin-2-yldisulfanyl)propan-1-ol
- 16-((3-((2-cyanobenzo[d]thiazol-6-yloxy)carbonyloxy)propyl)disulfanyl)hexadecanoic acid
- (S)-2-(6-((3-((15-carboxypentadecyl)disulfanyl)propoxy)carbonyloxy)benzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
- (S)-2-(6-((3-((15-carboxypentadecyl)disulfanyl)propoxy)carbonyloxy)-benzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
- methyl 16-((3-((2-cyanobenzo[d]thiazol-6-yloxy)carbonyloxy)propyl)disulfanyl)hexadecanoate
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
24.8 °F - closed cup
Flash Point(C)
-4 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service