12342
Iron(III) oxide
purified, ≥95%
Synonym(s):
Ferric oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Fe2O3
CAS Number:
Molecular Weight:
159.69
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Recommended Products
Assay
≥95%
form
solid
quality
purified
dilution
(for analytical testing)
loss
≤0.6% loss on ignition
SMILES string
O=[Fe]O[Fe]=O
InChI
1S/2Fe.3O
InChI key
JEIPFZHSYJVQDO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Iron is present in the form of iron(III) oxide in almost all natural sources of water. It is widely used as a supplement of iron and in solar photoelectrolysis cells. It is a highly stable n-type semiconductor, and possesses a band gap of 2.0 to 2.2eV. It is widely used in solar photoelectrolysis cells. Self-organized nanoporous iron(III) oxide (haematite) has been prepared.
Application
Iron(III) oxide may be used in the preparation of iron(III) oxide-hydrate gels.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The structure of iron (III) oxide-hydrate gels.
Van der Giessen AA.
J. Inorg. Nucl. Chem., 28(10), 2155-2159 (1966)
Light-induced reduction of natural iron (III) oxide and its relevance to phytoplankton.
Finden DAS, et al.
Nature, 309, 783-784 (1984)
Synthesis and photoelectrochemical properties of nanoporous iron (III) oxide by potentiostatic anodization.
Prakasam HE, et al.
Nanotechnology, 17(17), 4285-4285 (2006)
William B Ingler et al.
Journal of the American Chemical Society, 126(33), 10238-10239 (2004-08-19)
Stable zinc-doped iron(III) oxide thin films that exhibit p-type behavior were synthesized by spray pyrolytic deposition (SPD) on conducting indium-doped tin oxide-coated glass substrate. The highest photocurrent density of 1.1 mA/cm2 was observed at an illumination intensity of 40 mW/cm2
Alice Panariti et al.
Journal of biomedical nanotechnology, 9(9), 1556-1569 (2013-08-29)
Magnetic nanoparticles have emerged as important players in current research in modern medicine since they can be used in medicine for diagnosis and/or therapeutic treatment of diseases. Among many therapeutic applications of iron-based nanoparticles, drug delivery and photothermal therapy are
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service