Z802034
Hellma® liquid calibration standards
UV14, for testing the photometric accuracy
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
Recommended Products
grade
certified reference material
manufacturer/tradename
Hellma (667-UV14)
application(s)
food and beverages
petroleum
pharmaceutical
plastics
format
neat
Looking for similar products? Visit Product Comparison Guide
General description
Consisting of :
Perchloric acid reference filter (667-UV14, wavelength 235; 257; 313; 350 nm)
Perchloric acid reference filter (667-UV14, wavelength 235; 257; 313; 350 nm)
Application
Hellma® liquid calibration standards for checking the photometric accuracy of spectrophotometers.
Features and Benefits
- Accredited to DAkkS Deutsche Akkreditierungsstelle (National accreditation body for the Federal Republic of Germany)
- Traceable to NIST (National Institute of Standards and Technology)
- Assured Quality of the measurement results
- Proven accuracy and reliability
Other Notes
The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.
Legal Information
Hellma is a registered trademark of Hellma GmbH
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service