Z801291
Hellma® fluorescence cuvettes, cells with tubes Quartz/Duran®
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 10x10 mm ± 0.2 mm, chamber volume 3500 μL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
Suprasil® quartz
description
4 windows
manufacturer/tradename
Hellma 221.001-QS
Hellma
parameter
200-2500 nm spectral range
technique(s)
photometry: suitable
H × W × D
40 mm × 12.5 mm × 12.5 mm
chamber volume
3500 μL
pathlength
10x10 mm ± 0.2 mm
detection method
fluorometric
Looking for similar products? Visit Product Comparison Guide
General description
macro, tube diameter 8 mm, tube length 80 mm, for measurements at high and low temperature
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service