Skip to Content
Merck
All Photos(1)

Documents

Y0001785

Dienogest

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(17a)-17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-cyanomethyl-17-hydroxy-estra-4,9-dien-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO2
CAS Number:
Molecular Weight:
311.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dienogest

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@@]1(CC#N)CC[C@@]2([H])[C@]3([H])CCC4=CC(CCC4=C3CC[C@@]21C)=O

InChI

1S/C20H25NO2/c1-19-8-6-16-15-5-3-14(22)12-13(15)2-4-17(16)18(19)7-9-20(19,23)10-11-21/h12,17-18,23H,2-10H2,1H3/t17-,18+,19+,20-/m1/s1

InChI key

AZFLJNIPTRTECV-FUMNGEBKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dienogest EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Dienogest is a fourth-generation progestin, an orally active synthetic progestogen with approximately 10% of the relative affinity of progesterone, but has a very potent progestagenic effect in the endometrium with some antiandrogenic activity and without any androgenic, glucocorticoid or mineralocorticoid effects. It has been used alone or in combination as a contraceptive and to treat uterine fibroids. Dienogest inhibits ovulation primarily through peripheral action rather than by central action on gonadotrophin secretion.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service