Skip to Content
Merck
All Photos(1)

Documents

Y0001395

Voriconazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2R,3S-2-(2,4-Difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, UK-109496

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3N5O
CAS Number:
Molecular Weight:
349.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

voriconazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](c1ncncc1F)[C@](O)(Cn2cncn2)c3ccc(F)cc3F

InChI

1S/C16H14F3N5O/c1-10(15-14(19)5-20-7-22-15)16(25,6-24-9-21-8-23-24)12-3-2-11(17)4-13(12)18/h2-5,7-10,25H,6H2,1H3/t10-,16+/m0/s1

InChI key

BCEHBSKCWLPMDN-MGPLVRAMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Voriconazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Voriconazole is an antifungal used to treat serious fungal infections. Voriconazole inhibits ergosterol synthesis by inhibiting CYP450-dependent 14-α sterol demethylase resulting in a depletion of ergosterol in fungal cell membranes.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Muta. 2 - Repr. 2 - STOT RE 1 - STOT RE 2 Oral - STOT SE 2 Oral

Target Organs

Eyes, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martyna Mroczyńska et al.
Microorganisms, 8(2) (2020-02-26)
Triazoles are the only compounds used as antibiotics in both medicine and agriculture. The presence of triazoles in the environment can contribute to the acquisition of azole resistance among isolates of Aspergillus fumigatus. The objective of this study was to
Kim Vanstraelen et al.
Antimicrobial agents and chemotherapy, 58(11), 6782-6789 (2014-09-04)
Setting the adequate dose for voriconazole is challenging due to its variable pharmacokinetics. We investigated the impact of hypoalbuminemia (<35 g/liter) on voriconazole pharmacokinetics in adult intensive care unit (ICU) patients treated with voriconazole (20 samples in 13 patients) as
Atsuko Sunada et al.
Ophthalmology, 121(10), 2059-2065 (2014-06-02)
To evaluate the effectiveness of topical agents for the treatment of Acanthamoeba keratitis (AK). Laboratory research. Fifty-six Acanthamoeba isolates from 56 patients with clinically proven AK were studied. The effectiveness of 7 agents against Acanthamoeba cysts was determined in vitro.
Yongxuan Hu et al.
Mycopathologia, 179(1-2), 103-109 (2014-11-05)
Chromoblastomycosis, a chronic fungal infection of skin and subcutaneous tissue caused by dematiaceous fungi, is associated with low cure and high relapse rates. Among all factors affecting clinical outcome, etiological agents have an important position. In southern China, Fonsecaea pedrosoi
Chang Hyun Park et al.
Cornea, 33(9), 928-934 (2014-07-12)
The aim of this study was to evaluate the toxicity of intrastromally injected voriconazole on corneal endothelial cells in rabbits. In total, 32 eyes of 16 rabbits (8 eyes for each group) were divided into 4 groups according to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service