Y0001077
Iopromide impurity A
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
5-Amino-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-N-methylbenzene-1,3-dicarboxamide
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
iopromide
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
General description
Application
Packaging
Other Notes
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service