Skip to Content
Merck
All Photos(1)

Documents

Y0000889

Flucloxacillin impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(+)-6-Aminopenicillanic acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-APA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O3S
CAS Number:
Molecular Weight:
216.26
Beilstein:
15080
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxacillin; benzylpenicillin, amoxicillin, flucloxacillin

manufacturer/tradename

EDQM

mp

198-200 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O

InChI

1S/C8H12N2O3S/c1-8(2)4(7(12)13)10-5(11)3(9)6(10)14-8/h3-4,6H,9H2,1-2H3,(H,12,13)/t3-,4+,6-/m1/s1

InChI key

NGHVIOIJCVXTGV-ALEPSDHESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flucloxacillin impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Garrod lecture. The influence of 6-aminopenicillanic acid on antibiotic development.
G N Rolinson
The Journal of antimicrobial chemotherapy, 22(1), 5-14 (1988-07-01)
G N Rolinson et al.
International journal of antimicrobial agents, 29(1), 3-8 (2006-12-02)
This year (2007) marks the 50th anniversary of [corrected] discovery of 6-aminopenicillanic acid (6-APA), the precursor of all semi-synthetic penicillins [corrected] This review, by a scientist who played a major part in the discovery and a physician who participated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service