Skip to Content
Merck
All Photos(1)

Key Documents

W440201

Sigma-Aldrich

3-Octyl butyrate

natural (US), ≥98%

Synonym(s):

Octan-3-yl butanoate

Sign Into View Organizational & Contract Pricing

Select a Size

W440201-SAMPLE-K
CHF 53.40
100 G
CHF 107.00
1 KG
CHF 513.00

CHF 53.40


Estimated to ship on09 May 2025


Request a Bulk Order

Select a Size

Change View
W440201-SAMPLE-K
CHF 53.40
100 G
CHF 107.00
1 KG
CHF 513.00

About This Item

Empirical Formula (Hill Notation):
C12H24O2
CAS Number:
Molecular Weight:
200.32
FEMA Number:
4402
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
green; musty; citrus; fruity
grade:
Kosher
natural (US)
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

CHF 53.40


Estimated to ship on09 May 2025


Request a Bulk Order

grade

Kosher
natural (US)

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

Assay

≥98%

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; musty; citrus; fruity

SMILES string

O(C(CCCCC)CC)C(=O)CCC

InChI

1S/C12H24O2/c1-4-7-8-10-11(6-3)14-12(13)9-5-2/h11H,4-10H2,1-3H3

InChI key

USSKZVIXWTWCGK-UHFFFAOYSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service