W353600
Dimethyl disulfide
natural, ≥98%, FG
Synonym(s):
DMDS, Methyl disulfide
About This Item
Recommended Products
biological source
broccoli
sugar cane
Quality Level
grade
FG
Halal
Kosher
natural
Agency
meets purity specifications of JECFA
reg. compliance
EU Regulation 1334/2008 & 178/2002
vapor density
3.24 (vs air)
vapor pressure
22 mmHg ( 20 °C)
Assay
≥98%
autoignition temp.
>572 °F
expl. lim.
16 %
refractive index
n20/D 1.525 (lit.)
bp
109 °C (lit.)
mp
−85 °C (lit.)
density
1.046 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
cabbage; onion; vegetable; sulfurous
SMILES string
CSSC
InChI
1S/C2H6S2/c1-3-4-2/h1-2H3
InChI key
WQOXQRCZOLPYPM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1 Inhalation - STOT SE 3
Target Organs
Central nervous system, Upper respiratory tract
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
59.0 °F - closed cup
Flash Point(C)
15 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service