Skip to Content
Merck
All Photos(1)

Documents

T2655

Supelco

Testosterone-d3

98 atom % D, analytical standard, for drug analysis

Synonym(s):

17β-Hydroxyandrost-4-en-3-one-16,16,17-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19D3H25O2
CAS Number:
Molecular Weight:
291.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

98 atom % D

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+3

SMILES string

[2H]C1([2H])C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@@]1([2H])O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1/i6D2,17D

InChI key

MUMGGOZAMZWBJJ-HZRGXLBSSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael F Wangler et al.
PLoS genetics, 13(6), e1006825-e1006825 (2017-06-24)
Peroxisome biogenesis disorders (PBD) are a group of multi-system human diseases due to mutations in the PEX genes that are responsible for peroxisome assembly and function. These disorders lead to global defects in peroxisomal function and result in severe brain
Lindsey S Treviño et al.
Nature communications, 11(1), 2316-2316 (2020-05-10)
Our early-life environment has a profound influence on developing organs that impacts metabolic function and determines disease susceptibility across the life-course. Using a rat model for exposure to an endocrine disrupting chemical (EDC), we show that early-life chemical exposure causes
Guro Forsdahl et al.
Drug testing and analysis (2018-03-24)
In doping control analysis, the characterization of urinary steroid metabolites is of high interest for a targeted and long-term detection of prohibited anabolic androgenic steroids (AAS). In this work, the structure of a long-term metabolite of dehydrochloromethyltestosterone (DHCMT) was elucidated.
Hyeung Kim et al.
Cell discovery, 3, 17034-17034 (2017-09-29)
CRISPR/Cas9 technology enables efficient loss-of-function analysis of human genes using somatic cells. Studies of essential genes, however, require conditional knockout (KO) cells. Here, we describe the generation of inducible CRISPR KO human cell lines for the subunits of the telosome/shelterin
Claudio Oliviero et al.
Theriogenology, 85(3), 476-482 (2015-10-18)
Boar stations raise only entire male pigs for selection as reproductive boars, but the majority of them will fail the selection process, ending at slaughter with a high risk of boar tainted meat. The aim of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service