Skip to Content
Merck
All Photos(1)

Documents

S114

Supelco

Nor-R-(+)-SCH-23390 hydrochloride

analytical standard

Synonym(s):

(R)-(+)-7-Chloro-8-hydroxy-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClNO · HCl
CAS Number:
Molecular Weight:
310.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.Oc1cc2[C@H](CNCCc2cc1Cl)c3ccccc3

InChI

1S/C16H16ClNO.ClH/c17-15-8-12-6-7-18-10-14(13(12)9-16(15)19)11-4-2-1-3-5-11;/h1-5,8-9,14,18-19H,6-7,10H2;1H/t14-;/m1./s1

InChI key

KTHOTRUZLPTFOY-PFEQFJNWSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service