PSRI02K
Refractive Index Standard at 20 °C 25 °C and 30 °C
Nominal RI Value 1.3891 at 25 °C, in 5 x 10mL Tamper-Evident Glass Vials
Select a Size
About This Item
Recommended Products
quality
in 5 x 10mL Tamper-Evident Glass Vials
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | PSRI09K | PSRI08K | PSRI07K |
---|---|---|---|
manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd |
quality in 5 x 10mL Tamper-Evident Glass Vials | quality in 5 x 10mL Tamper-Evident Glass Vials | quality in 5 x 10mL Tamper-Evident Glass Vials | quality in 5 x 10mL Tamper-Evident Glass Vials |
shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
10.4 °F - closed cup
Flash Point(C)
-12 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service