Skip to Content
Merck
All Photos(1)

Key Documents

PHR2990

Supelco

Alosetron Hydrochloride

pharmaceutical secondary standard, certified reference material

Synonym(s):

Alosetron hydrochloride, 2,3,4,5-Tetrahydro-5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-1H-pyrido[4,3-b]indol-1-one hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 520.00

CHF 520.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
500 MG
CHF 520.00

About This Item

Empirical Formula (Hill Notation):
C17H18N4O·HCl
CAS Number:
Molecular Weight:
330.81
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CHF 520.00


Estimated to ship on27 April 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1013807

API family

alosetron

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

CN1C2=C(C=CC=C2)C3=C1CCN(CC4=C(C)N=CN4)C3=O.Cl

InChI

1S/C17H18N4O.ClH/c1-11-13(19-10-18-11)9-21-8-7-15-16(17(21)22)12-5-3-4-6-14(12)20(15)2;/h3-6,10H,7-9H2,1-2H3,(H,18,19);1H

InChI key

FNYQZOVOVDSGJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Alosetron is a potent and highly selective antagonist of serotonin 5-HT3 receptors, nonselective cation channels found predominantly in the enteric nervous system of the gastrointestinal tract. These receptors are involved in the regulation of visceral pain, colonic transit and GI secretions that can contribute to the pathophysiology of irritable bowel syndrome (IBS). Alosetron is used clinically for treatment of women with severe diarrhea-predominant IBS.
Alosetron is a potent and highly selective antagonist.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service