Skip to Content
Merck
All Photos(1)

Documents

PHR2932

Supelco

Rosiglitazone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Rosiglitazone, 5-[[4-[2-(Methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H19N3O3S
CAS Number:
Molecular Weight:
357.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1605817

API family

rosiglitazone

form

solid

packaging

pkg of 300 mg

application(s)

pharmaceutical small molecule

storage temp.

2-8°C

SMILES string

CN(CCOc1ccc(CC2SC(=O)NC2=O)cc1)c3ccccn3

InChI

1S/C18H19N3O3S/c1-21(16-4-2-3-9-19-16)10-11-24-14-7-5-13(6-8-14)12-15-17(22)20-18(23)25-15/h2-9,15H,10-12H2,1H3,(H,20,22,23)

InChI key

YASAKCUCGLMORW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Rosiglitazone is a potent agonist for PPARγ with an EC50 of 43 nM for the human receptor. It is antidiabetic, working as an insulin sensitizer by binding to the PPARγ receptors in fat cells and making the cells more responsive to insulin.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service