Skip to Content
Merck
All Photos(1)

Documents

NIST460

Beryllium-copper alloy

NIST® SRM® 460

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is in the form of chips sized between 0.05 and 1.18 mm sieve openings (35 and 16 mesh). It is intended primarily for use in chemical methods of analysis. For more information, plese refer to the SDS and COA.

SRM 460_cert

SRM 460_SDS

Other Notes

Certified for the analytes listed below.
Beryllium (Be), Cobalt (Co), Nickel (Ni), Iron (Fe), Silicon (Si), Aluminium (Al), Tin (Sn), Zinc (Zn), Chromium (Cr), Lead (Pb), Magnesium (Mg)

Reference/Informational value is provided for the following analytes.
Antimony (Sb), Manganese (Mn), Silver (Ag), Sulfur (S), Titanium (Ti), Zirconium (Zr), Copper (Cu)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service