Skip to Content
Merck
All Photos(1)

Key Documents

NIST1848

Lubricating oil additive package

NIST® SRM® 1848

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 100 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in the evaluation of methods and the calibration of equipment used in the analysis of lubricating oil additive packages, engine lubricating oils, and materials of a similar matrix. SRM 1848 consists of a typical additive package used in the manufacture of crankcase lubricating oil for gasoline engines. A unit of SRM 1848 consists of an amber, borosilicate glass bottle containing approximately 100 g of material. For more information, please refer to the COA and SDS.

SRM 1848_cert

SRM 1848_SDS

Other Notes

Certified for the analytes listed below.
Boron (B), Calcium (Ca), Chlorine (Cl), Magnesium (Mg), Phosphorus (P), Sulfur (S), Zinc (Zn)

Reference/Informational value is provided for the following analytes. Hydrogen (H), Nitrogen (N), Silicon (Si)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

343.4 °F

Flash Point(C)

173 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service