Skip to Content
Merck
All Photos(1)

Documents

NIST16F

Basic open-hearth steel

NIST® SRM® 16f, 1% carbon

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 150 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in evaluating chemical and instrumental methods of analysis. A unit of SRM 16f consists of a bottle containing approximately 150 g of chips sized between 0.50 mm and 1.18 mm sieve openings (35 mesh and 16 mesh). For more information, please refer to the SDS and COA.

SRM 16F_cert

SRM 16F_SDS

Other Notes

Certified for the analytes listed below.
Carbon (C), Chromium (Cr), Cobalt (Co), Copper (Cu), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Phosphorus (P), Silicon (Si) Sulfur (S), Vanadium (V)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service