Skip to Content
Merck
All Photos(1)

Documents

NIST1598A

Inorganic constituents in animal serum

NIST® SRM® 1598a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

packaging

pkg of 2 vials

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical testing

format

matrix material

General description

This Standard Reference Material (SRM) is a serum sample derived from a mixture of serum from healthy bovine and porcine animals collected under strict protocols designed to preserve the original composition and minimize contamination. A unit of SRM 1598a consists of two capped polyethylene vials, each containing 5 mL to 6 mL of frozen serum. For more information, please refer to the SDS and COA.

SRM 1598A_cert

SRM 1598A _SDS

Application

The inorganic constituents in animal serum reference standard (RM) are intended primarily for use in calibrating instrumentation and evaluating the accuracy of analytical methods for selected elements in blood serum, plasma, and similar biological fluids.

Other Notes

  • Expiration details and storage and use instructions are provided on the NIST certificate
  • Notes on biomaterial handling, disposal, and transport documented in MSDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service