N0800000
Nikethamide
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
N,N-Diethylnicotinamide, Nicotinic acid diethylamide, ‘Nikethamide’
About This Item
grade
pharmaceutical primary standard
API family
nikethamide
manufacturer/tradename
EDQM
refractive index
n20/D 1.524 (lit.)
bp
296-300 °C (lit.)
mp
23 °C (lit.)
density
1.06 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
CCN(CC)C(=O)c1cccnc1
InChI
1S/C10H14N2O/c1-3-12(4-2)10(13)9-6-5-7-11-8-9/h5-8H,3-4H2,1-2H3
InChI key
NCYVXEGFNDZQCU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service