Skip to Content
Merck
All Photos(1)

Key Documents

N0025000

Nadroparin calcium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Heparin calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nadroparin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nadroparin calcium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Andreozzi et al.
Minerva cardioangiologica, 48(12 Suppl 1), 9-14 (2001-03-20)
Thrombophlebitis of the superficial veins (SVT) of the leg is usually regarded as a mild and uncomplicated disease. Although this is generally true for acute thrombosis of the branches of the saphenous vein, the natural history of SVT involving the
C E Geffroy et al.
Presse medicale (Paris, France : 1983), 31(7), 303-311 (2002-03-20)
To control whether prescriptions of curative doses of heparin (non fractioned heparins, enoxaparin, tinzaparin) in the hospital complied with the official recommendations; to provide the physicians with information adapted to the recorded misuse and to evaluate the influence of this
Gabriel Lima-Oliveira et al.
Clinical biochemistry, 45(9), 683-687 (2012-03-24)
Evaluate whether four different brands/types of heparin coated syringes can represent a source of variability in blood gas analysis (BGA). Blood was collected from one hundred volunteers into different syringes: Syringe I (lithium heparin and calcium balanced); Syringe II: in-house
Cutaneous allergic reaction to heparins: subcutaneous but not intravenous provocation.
Maria Maroto-Iitani et al.
Contact dermatitis, 52(4), 228-230 (2005-04-30)
T Ripert et al.
Transplantation proceedings, 41(10), 4193-4196 (2009-12-17)
Renal allograft vascular thrombosis is a complication that often results in graft loss. Since there are no guidelines on immediate postoperative thromboprophylaxis, we performed a telephone survey of clinical practice in all renal transplantation centers in France. Each center considered

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service