Skip to Content
Merck
All Photos(1)

Key Documents

IRMM654

Zinc 68Zn solution (certified for isotope abundance ratio)

IRMM®, certified reference material, 0.5 M in nitric acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CHF 856.00

CHF 856.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
CHF 856.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CHF 856.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

IRMM®

isotopic purity

 68Zn

manufacturer/tradename

JRC

concentration

0.5 M in nitric acid

format

single component solution

storage temp.

10-30°C

Analysis Note

For more information please see:
IRMM654

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service