Skip to Content
Merck
All Photos(1)

Key Documents

IRMM427

Pike-perch (PFASs in fish tissue)

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 340.00

CHF 340.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CHF 340.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CHF 340.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

IRMM®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

JRC

matrix

fish

application(s)

PFAS testing
food and beverages

format

matrix material

Analysis Note

For more information please see:
IRMM427

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants: Branched perfluorooctane sulfonate (br-PFOS), Linear perfluorooctane sulfonate (L-PFOS), Perfluoroctane sulfonamide (FOSA), Perfluorodecanoic acid (PFDA), Perfluorododecanoic acid, Perfluorohexanesulfonate (PFHxS), Perfluorononanoic acid, Perfluorotetradecanoic acid (PFTeDA), Perfluorotridecanoic acid (PFTrDA), Perfluoroundecanoic acid, Total perfluorooctane sulfonate (tot-PFOS)

Matrix Group: Fish and Seafood

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the mass of this sample? Is it an entire fish, or a sample of fish tissue? If a tissue sample, what type of tissue? Are documents with known sample PFAS concentrations provided with this sample? Thank you!

    1 answer
    1. The CRM is available in glass jars containing approximately 35 g of fish paste. The minimum amount of sample to be used is 1 g.

      The base material employed for CRM IRMM-427 was pike-perch (Lucioperca lucioperca)
      fillets originating from the rivers Nieuwe Merwede and Amer in The Netherlands. The fish
      was selected after pre-screening experiments conducted by LC-MS/MS on different naturally
      contaminated species where the presence of a number of PFASs was investigated.

      Please view the following link for more information, documentation, and PFAS recorded concentrations for this product:
      https://crm.jrc.ec.europa.eu/p/q/IRMM-427/IRMM-427-PIKE-PERCH/IRMM-427

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service