Skip to Content
Merck
All Photos(1)

Documents

HLA002

Sigma-Aldrich

Jurkat HLA Panel Cell Lines

Synonym(s):

CAR-T, Checkpoint Inhibitors, Immuno-oncology, Immunotherapy, Oncology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human (T Lymphocyte (blood))

growth mode

suspension

shipped in

dry ice

storage temp.

−196°C

General description

The STR profile of these cell lines match that of its parental cell line - ECACC catalog number 88042803.

This product is a panel of eleven (11) genetically modified cell lines targeting MHC Class I HLA molecules in a Jurkat T lymphocyte parental cell line background.  The full HLA panel consists of a Beta-2 microglobulin (β2M) knockout cell line and an additional ten (10) monoallelic HLA expression cell lines.  

CompoZr® Zinc Finger Nuclease (ZFN) technology was used to generate the β2M knockout cell line, which does not express endogenous cell surface MHC Class I HLA molecules (HLA-A or HLA-B) as measured by fluorescence-activated cell sorting (FACS).  

MISSION® lentivirus was used to generate the ten (10)  monoallelic HLA cell lines, which express individual HLA-A or HLA-B subtypes on the cell surface via a β2M:HLA fusion protein as described in Nature Biotechnology 35, 765-772, (2017) and as demonstrated via FACS analysis.  

Cell Line Description

Derived from Jurkat FHCRC. An IL-2 producing cell line, derived by incubating the cells at 41°C for 48 hours followed by a limiting dilution cloning over macrophages.

Features and Benefits

Unlike wild type Jurkat cells, which express multiple HLA subtypes, the Jurkat HLA panel cell lines express individual MHC Class I HLA subtypes. These monoallelic HLA cell lines can be used to study peptide binding and presentation in the context of individual HLA subtypes. The panel can also be used to develop cell-based assays to evaluate candidate cancer immunotherapies and identify tumor-specific neoantigens.

Components

HLA002A-1VL: Jurkat B2M KO Cells
HLA002B-1VL: Jurkat HLA-A*02:01 Cells
HLA002C-1VL: Jurkat HLA-A*01:01 Cells
HLA002D-1VL: Jurkat HLA-A*03:01 Cells
HLA002E-1VL: Jurkat HLA-A*11:01 Cells
HLA002F-1VL: Jurkat HLA-A*24:02 Cells
HLA002G-1VL: Jurkat HLA-B*15:10 Cells
HLA002H-1VL: Jurkat HLA-B*07:02 Cells
HLA002I-1VL: Jurkat HLA-B*08:01 Cells
HLA002J-1VL: Jurkat HLA-B*35:01 Cells
HLA002K-1VL: Jurkat HLA-B*40:01 Cells

Quality

Tested for Mycoplasma, bacterial and fungal content, post-freeze viability, and short terminal repeat (STR) analysis for cell line identification.

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service