Skip to Content
Merck
All Photos(1)

Key Documents

F0385

Sigma-Aldrich

Minimum Essential Medium Eagle

With Earle′s salts, without ʟ-glutamine, phenol red, and sodim bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
CHF 31.90

CHF 31.90


Estimated to ship on11 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
CHF 31.90

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

CHF 31.90


Estimated to ship on11 May 2025


Request a Bulk Order

Product Name

Minimum Essential Medium Eagle, With Earle′s salts, without L-glutamine, phenol red, and sodim bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

2-8°C

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service