Skip to Content
Merck
All Photos(1)

Key Documents

ERMDA483IFCC

anti-PR3 IgG in human serum

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 211.00

CHF 211.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CHF 211.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CHF 211.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

clinical testing

format

matrix material

storage temp.

−70°C

General description

A serum protein reference material intended for the standardisation of measurements of immunoglobulin G proteinase 3 anti-neutrophil cytoplasmic autoantibodies (IgG PR3 ANCA). The material was produced according to ISO Guide 34:2009 and is certified in accordance with ISO Guide 35:2006

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service