Skip to Content
Merck
All Photos(1)

Key Documents

ERMCC580

Estuarine sediment (total Hg and methylmercury)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

40 G
CHF 432.00

CHF 432.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
40 G
CHF 432.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CHF 432.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®
certified by the BSC

composition

CH3Hg+, 0.0755 mg/kg ±0.0037 mg/kg by
total Hg, 132 mg/kg ±3 mg/kg by

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

storage temp.

2-8°C

Packaging

Glass bottle containing ~40 g powder

Analysis Note

For more information please see:
ERMCC580

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service