Skip to Content
Merck
All Photos(1)

Key Documents

ERMBB446

Pork fat (PCB)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 242.00

CHF 242.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
CHF 242.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

CHF 242.00


Estimated to ship on20 May 2025


Request a Bulk Order

grade

certified reference material

Agency

ERM®
certified by the BSC

description

high level

manufacturer/tradename

JRC

format

matrix material

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMBB446

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic pollutants: BDE 47, Lindane, PCB 101, PCB 118, PCB 138, PCB 153, PCB 180, PCB 28, PCB 52

Matrix Group: Meat

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service