Skip to Content
Merck
All Photos(1)

Key Documents

ERMAD623

BCR-ABL pDNA calibrant

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 0.6 ML
CHF 148.00

CHF 148.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
6 X 0.6 ML
CHF 148.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CHF 148.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

general analytical
genomic analysis

format

single component solution

storage temp.

−20°C

General description

Certified for the numbers of specific DNA fragments per plasmid (BCR-ABL b3a2 transcript, BCR transcript and GUSB transcript) and the copy number concentration of the plasmid (ERM-AD 623a, ERM-AD 623b, ERM-AD 623c, ERM-AD 623d, ERM-AD 623e, ERM-AD 623f)

Application

BCR-ABL pDNA calibrant has been used as a calibration reference material for evaluating the performance of the duplex BCR-ABL1 and ABL1 assay by reverse transcription–quantitative PCR (RT-qPCR).[1]

Analysis Note

For more information please see:
ERMAD623

Legal Information

ERM is a registered trademark of European Commission

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RT-qPCR and RT-digital PCR: a comparison of different platforms for the evaluation of residual disease in chronic myeloid leukemia
Alikian M, et al.
Clinical Chemistry, 63, 525-531 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service