Skip to Content
Merck
All Photos(1)

Key Documents

E2377

Sigma-Aldrich

Eriochrome® Black T

suitable for metal indicator

Synonym(s):

Mordant Black 11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12N3NaO7S
CAS Number:
Molecular Weight:
461.38
Colour Index Number:
14645
Beilstein:
4121162
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, ≥60%

technique(s)

titration: suitable

suitability

suitable for metal indicator

SMILES string

[Na+].Oc1cc(c2cc(ccc2c1\N=N\c3ccc4ccccc4c3O)[N+]([O-])=O)S([O-])(=O)=O

InChI

1S/C20H13N3O7S.Na/c24-17-10-18(31(28,29)30)15-9-12(23(26)27)6-7-14(15)19(17)22-21-16-8-5-11-3-1-2-4-13(11)20(16)25;/h1-10,24-25H,(H,28,29,30);/q;+1/p-1/b22-21+;

InChI key

AMMWFYKTZVIRFN-QUABFQRHSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eriochrome is a registered trademark of Huntsman Petrochemical, LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kvernmo et al.
The international journal of biochemistry & cell biology, 28(3), 303-309 (1996-03-01)
Due to the central role of sorbitol dehydrogenase in diabetic cataract, it is important to examine this enzyme's interaction with different inhibitory compounds such as dyes. The aim of the study was to investigate the binding of Cibacron Blue and
Hong Yao et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 23(6), 677-682 (2007-06-19)
An electropolymerized film of eriochrome black T (EBT) has been prepared at a glassy carbon electrode (GCE) by cyclic voltammetry (CV). The poly(EBT) membrane at GCE exhibits an excellent electrocatalytic activity towards the oxidation of epinephrine (EP), ascorbic acid (AA)
J L Seltzer et al.
Collagen and related research, 7(6), 399-407 (1987-12-01)
The intracellular degradation of interstitial collagen is accomplished by two neutral metalloproteases, collagenase and gelatinase. Both enzymes are inhibited by metal chelating agents, by certain sulfhydryl reagents, and by similar protein inhibitors. Here, we demonstrate that the dye eriochrome black
Liqing Lin et al.
Talanta, 80(5), 2113-2119 (2010-02-16)
A novel electrochemical biosensor is described for detection of breakpoint cluster region gene and a cellular abl (BCR/ABL) fusion gene in chronic myelogenous leukemia (CML) by using thiolated-hairpin locked nucleic acids (LNA) as the capture probe. The hairpin LNA probe
Li-Tai Jin et al.
Proteomics, 6(8), 2334-2337 (2006-02-24)
A quick, sensitive, and MALDI-TOF MS compatible silver staining method, namely Eriochrome black T (EBT)-silver method, is described. The method can detect 0.05-0.2 ng protein within 60 min in SDS-PAGE gels. EBT dye was used as a silver ion sensitizer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service