Skip to Content
Merck
All Photos(1)

Key Documents

CRMUTAKR

TAG Flash Point, Jet Aviation Fuel

UKAS ISO Guide 34 Certified (40.4°C)

Synonym(s):

Kerosine (petroleum), sweetened

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
CHF 490.00

CHF 490.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
CHF 490.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

CHF 490.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

packaging

pkg of 250 mL

manufacturer/tradename

Paragon Scientific Ltd

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 - 158.0 °F - closed cup

Flash Point(C)

29 - 70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service